1-Amino-1H-pyrrole-2-carboxamide ≥96%

Supplier: Matrix Scientific

Synonyms: 1-Aminopyrrole-2-carboxamide

070161-500MG 070161-1G 070161-5G
101943-598EA 815.38 USD
101943-598 101943-600 101943-602
1-Amino-1H-pyrrole-2-carboxamide ≥96%
1-Amino-1H-pyrrole-2-carboxamide

Hazard: Irritant

Formula: C₅H₇N₃O
MW: 125.13 g/mol
MDL Number: MFCD11053506
CAS Number: 159326-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR