6-Bromo-3,4-dihydro-2H-S,S-dioxothiochromen-4-amine hydrochloride ≥96%

Supplier: Matrix Scientific

Synonyms: 4-Amino-6-bromothiochroman 1,1-dioxide hydrochloride

070159-500MG 070159-1G 070159-5G
101943-586EA 1221.54 USD
101943-586 101943-588 101943-590
6-Bromo-3,4-dihydro-2H-S,S-dioxothiochromen-4-amine hydrochloride ≥96%
6-Bromo-3,4-dihydro-2H-S,S-dioxothiochromen-4-amine hydrochloride

Hazard: Irritant

Formula: C₉H₁₁BrClNO₂S
MW: 312.62 g/mol
MDL Number: MFCD11616744
CAS Number: 1172986-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR