4-(1-Pyrrolidinylcarbonyl)phenylboronic acid ≥96%

Supplier: Matrix Scientific

Synonyms: 4-(1-Pyrrolidinylcarbonyl)benzeneboronic acid, 4-(Pyrrolidine-1-carbonyl)phenylboronic acid, (4-(Pyrrolidine-1-carbonyl)phenyl)boronic acid

070157-1G 070157-5G 070157-25G
101943-574EA 140 USD
101943-574 101943-576 101943-578
4-(1-Pyrrolidinylcarbonyl)phenylboronic acid ≥96%
4-(1-Pyrrolidinylcarbonyl)phenylboronic acid

Hazard: Irritant

Formula: C₁₁H₁₄BNO₃
MW: 219.05 g/mol
Melting Pt: 136-140°C
MDL Number: MFCD03411950
CAS Number: 389621-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR