4-Hydrazino-1H-pyrimidin-2-one ≥95%

Supplier: Matrix Scientific

070156-1G 070156-5G
101943-570EA 815.38 USD
101943-570 101943-572
4-Hydrazino-1H-pyrimidin-2-one ≥95%
4-Hydrazino-1H-pyrimidin-2-one

Hazard: Irritant

Formula: C₄H₆N₄O
MDL Number: MFCD01749722
CAS Number: 3310-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR