5-Bromo-4-chloroquinazoline ≥97%

Supplier: Matrix Scientific

070154-1G 070154-5G
101943-562EA 360 USD
101943-562 101943-564
5-Bromo-4-chloroquinazoline ≥97%
5-Bromo-4-chloroquinazoline

Hazard: Irritant

Formula: C₈H₄BrClN₂
MW: 243.487 g/mol
MDL Number: MFCD09739114
CAS Number: 2148-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR