2-Acetylaminothiazole-5-carboxylicacid ethyl ester ≥98%

Supplier: Matrix Scientific

070152-1G 070152-5G
101943-552EA 175.38 USD
101943-552 101943-554
2-Acetylaminothiazole-5-carboxylicacid ethyl ester ≥98%
2-Acetylaminothiazole-5-carboxylicacid ethyl ester

Hazard: Irritant

Formula: C₈H₁₀N₂O₃S
MDL Number: MFCD09909304
CAS Number: 106840-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR