3-Bromo-5,6,7,8-tetrahydro-1,6-naphthyridine hydrochloride ≥95%

Supplier: Matrix Scientific

070150-1G 070150-5G
101943-544EA 1338.46 USD
101943-544 101943-546
3-Bromo-5,6,7,8-tetrahydro-1,6-naphthyridine hydrochloride ≥95%
3-Bromo-5,6,7,8-tetrahydro-1,6-naphthyridine hydrochloride

Hazard: Irritant

Formula: C₈H₁₀BrClN₂
MDL Number: MFCD09701301
CAS Number: 1159010-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR