Methyl-8-Bromo-6-methylimidazo[1,2-a]pyridine-2-carboxylate ≥97%

Supplier: Matrix Scientific

070147-500MG 070147-1G
101943-530EA 553.85 USD
101943-530 101943-532
Methyl-8-Bromo-6-methylimidazo[1,2-a]pyridine-2-carboxylate ≥97%
Methyl-8-Bromo-6-methylimidazo[1,2-a]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₉BrN₂O₂
MDL Number: MFCD12026326
CAS Number: 1171424-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR