6-Bromo-7-methyl[1,2,4]triazolo[1,5-a]pyridine ≥95%

Supplier: Matrix Scientific

070145-500MG 070145-1G
101943-522EA 684.62 USD
101943-522 101943-524
6-Bromo-7-methyl[1,2,4]triazolo[1,5-a]pyridine ≥95%
6-Bromo-7-methyl[1,2,4]triazolo[1,5-a]pyridine

Hazard: Irritant

Formula: C₇H₆BrN₃
MDL Number: MFCD12026325
CAS Number: 1172534-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR