6-Bromo-8-methyl[1,2,4]triazolo[1,5-a]pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-8-methyl-[1,2,4]triazolo[1,5-a]pyridine

070144-500MG 070144-1G
101943-518EA 760 USD
101943-518 101943-520
6-Bromo-8-methyl[1,2,4]triazolo[1,5-a]pyridine ≥95%
6-Bromo-8-methyl[1,2,4]triazolo[1,5-a]pyridine

Hazard: Irritant

Formula: C₇H₆BrN₃
MW: 212.05 g/mol
MDL Number: MFCD09038166
CAS Number: 899429-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR