7-Bromo-2-chloromethylimidazo[1,2-a]pyridine ≥96%

Supplier: Matrix Scientific

070143-500MG 070143-1G
101943-514EA 1978.46 USD
101943-514 101943-516
7-Bromo-2-chloromethylimidazo[1,2-a]pyridine ≥96%
7-Bromo-2-chloromethylimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₈H₆BrClN₂
MDL Number: MFCD09995266
CAS Number: 1019023-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR