7-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid methyl ester ≥98%

Supplier: Matrix Scientific

Synonyms: Methyl 7-bromoimidazo[1,2-a]pyridine-2-carboxylate

070141-1G 070141-5G
101943-506EA 1106.15 USD
101943-506 101943-508
7-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid methyl ester ≥98%
7-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid methyl ester

Hazard: Irritant

Formula: C₉H₇BrN₂O₂
MW: 255.08 g/mol
MDL Number: MFCD12026323
CAS Number: 1170024-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR