2-Dimethylaminomethylenecyclohexane-1,3-dione ≥97%

Supplier: Matrix Scientific

070139-1G 070139-5G
101943-496EA 349.23 USD
101943-496 101943-498
2-Dimethylaminomethylenecyclohexane-1,3-dione ≥97%
2-Dimethylaminomethylenecyclohexane-1,3-dione

Hazard: Irritant

Formula: C₉H₁₃NO₂
MW: 167.21 g/mol
MDL Number: MFCD00172118
CAS Number: 85302-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR