1H-Indazole-4-boronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: (1H-Indazol-4-yl)boronic acid, 1H-indazole-4-boronic acid

070138-500MG 070138-1G 070138-5G
101943-490EA 107.69 USD
101943-490 101943-492 101943-494
1H-Indazole-4-boronic acid ≥97%
1H-Indazole-4-boronic acid

Hazard: Irritant

Formula: C₇H₇BN₂O₂
MW: 161.9500 g/mol
MDL Number: MFCD09878901
CAS Number: 1023595-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR