2-Pyridineboronic acid dimethyl ester ≥97%
Supplier: Matrix Scientific
070137-1G
070137-5G
101943-486EA
569.23
USD
101943-486
101943-488
2-Pyridineboronic acid dimethyl ester ≥97%
2-Pyridineboronic acid dimethyl ester
Hazard: Irritant
Formula:
C₇H₁₀BNO₂ MW: 150.97 |
MDL Number:
MFCD03412109 CAS Number: 136805-54-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...