3-Bromo-5-[1,3]dioxolan-2-yl-4,5-dihydroisoxazole ≥95%

Supplier: Matrix Scientific

070136-1G 070136-5G
101943-482EA 1396.92 USD
101943-482 101943-484
3-Bromo-5-[1,3]dioxolan-2-yl-4,5-dihydroisoxazole ≥95%
3-Bromo-5-[1,3]dioxolan-2-yl-4,5-dihydroisoxazole

Hazard: Irritant

Formula: C₆H₈BrNO₃
MDL Number: MFCD11707007
CAS Number: 1120215-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR