2-(3-Bromo-4,5-dihydroisoxazol-5-yl)pyridine ≥95%

Supplier: Matrix Scientific

070135-1G 070135-5G
101943-478EA 1221.54 USD
101943-478 101943-480
2-(3-Bromo-4,5-dihydroisoxazol-5-yl)pyridine ≥95%
2-(3-Bromo-4,5-dihydroisoxazol-5-yl)pyridine

Hazard: Irritant

Formula: C₈H₇BrN₂O
MDL Number: MFCD11707006
CAS Number: 1120215-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR