3-Bromo-5-(4-methoxyphenyl)-4,5-dihydroisoxazole ≥95%

Supplier: Matrix Scientific

070133-1G 070133-5G
101943-470EA 1047.69 USD
101943-470 101943-472
3-Bromo-5-(4-methoxyphenyl)-4,5-dihydroisoxazole ≥95%
3-Bromo-5-(4-methoxyphenyl)-4,5-dihydroisoxazole

Hazard: Irritant

Formula: C₁₀H₁₀BrNO₂
MDL Number: MFCD11707004
CAS Number: 1120215-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR