3-Bromo-5-phenyl-4,5-dihydroisoxazole ≥93%

Supplier: Matrix Scientific

070132-1G 070132-5G
101943-466EA 1047.69 USD
101943-466 101943-468
3-Bromo-5-phenyl-4,5-dihydroisoxazole ≥93%
3-Bromo-5-phenyl-4,5-dihydroisoxazole

Hazard: Irritant

Formula: C₉H₈BrNO
MDL Number: MFCD11707003
CAS Number: 86256-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR