4-Methyl-3-(tributylstannyl)pyridine ≥96%
Supplier: Matrix Scientific
070129-500MG
070129-1G
070129-5G
101943-452EA
349.23
USD
101943-452
101943-454
101943-456
4-Methyl-3-(tributylstannyl)pyridine ≥96%
4-Methyl-3-(tributylstannyl)pyridine
Hazard: Irritant
Formula:
C₁₈H₃₃NSn |
MDL Number:
MFCD11707000 CAS Number: 1204580-81-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...