2-(Trifluoromethyl)-1,3-bis(dimethylamino)trimethinium hexafluorophosphate ≥97%

Supplier: Matrix Scientific

070128-1G 070128-5G
101943-448EA 524.62 USD
101943-448 101943-450
2-(Trifluoromethyl)-1,3-bis(dimethylamino)trimethinium hexafluorophosphate ≥97%
2-(Trifluoromethyl)-1,3-bis(dimethylamino)trimethinium hexafluorophosphate

Hazard: Irritant

Formula: C₈H₁₄F₉N₂P
MDL Number: MFCD08461601
CAS Number: 291756-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR