Methyl pyrazolo[1,5-a]pyridine-3-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Pyrazolo[1,5-a]pyridine-3-carboxylic acid methyl ester

070127-500MG 070127-1G 070127-5G
101943-442EA 233.85 USD
101943-442 101943-444 101943-446
Methyl pyrazolo[1,5-a]pyridine-3-carboxylate ≥97%
Methyl pyrazolo[1,5-a]pyridine-3-carboxylate

Hazard: Irritant

Formula: C₉H₈N₂O₂
MDL Number: MFCD09752651
CAS Number: 63237-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR