(3S,4S)-3-Bromo-4-hydroxydihydrofuran-2-one ≥96%

Supplier: Matrix Scientific

070126-500MG 070126-1G
101943-438EA 1106.15 USD
101943-438 101943-440
(3S,4S)-3-Bromo-4-hydroxydihydrofuran-2-one ≥96%
(3S,4S)-3-Bromo-4-hydroxydihydrofuran-2-one

Hazard: Irritant

Formula: C₄H₅BrO₃
MDL Number: MFCD11706998
CAS Number: 117858-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR