1-Benzotriazol-1-yl-3-chloropropan-2-one ≥90%

Supplier: Matrix Scientific

070124-1G 070124-5G
101943-430EA 263.08 USD
101943-430 101943-432
1-Benzotriazol-1-yl-3-chloropropan-2-one ≥90%
1-Benzotriazol-1-yl-3-chloropropan-2-one

Hazard: Irritant

Formula: C₉H₈ClN₃O
MDL Number: MFCD01823137
CAS Number: 305851-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR