Ethyl-6,7-dichloro-3-hydroxyquinoxaline-2-carboxylate ≥97%

Supplier: Matrix Scientific

070103-500MG 070103-5G
101943-338EA 640 USD
101943-338 101943-342
Ethyl-6,7-dichloro-3-hydroxyquinoxaline-2-carboxylate ≥97%
Ethyl-6,7-dichloro-3-hydroxyquinoxaline-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₈Cl₂N₂O₃
MDL Number: MFCD11045110
CAS Number: 60578-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR