6-Bromoindazole-1-carboxylic acid tert-butyl ester ≥98%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 6-bromo-1H-indazole-1-carboxylate, 6-Bromo-1H-indazole, N1-BOC protected
070087-500MG
070087-5G
101943-264EA
524.62
USD
101943-264
101943-268
6-Bromoindazole-1-carboxylic acid tert-butyl ester ≥98%
6-Bromoindazole-1-carboxylic acid tert-butyl ester
Hazard: Irritant
Formula:
C₁₂H₁₃BrN₂O₂ MW: 297.16 g/mol |
MDL Number:
MFCD06739133 CAS Number: 877264-77-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...