1H-Indazole-5-boronic acid ≥97%

Supplier: Matrix Scientific

070068-500MG 070068-5G
101943-172EA 130.77 USD
101943-172 101943-176
1H-Indazole-5-boronic acid ≥97%
1H-Indazole-5-boronic acid

Hazard: Irritant

Formula: C₇H₇BN₂O₂
MW: 161.9537 g/mol
MDL Number: MFCD06739151
CAS Number: 338454-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR