3-Bromo-1H-pyrazolo[4,3-c]pyridine ≥98%

Supplier: Matrix Scientific

070066-250MG 070066-1G
101943-160EA 640 USD
101943-160 101943-164
3-Bromo-1H-pyrazolo[4,3-c]pyridine ≥98%
3-Bromo-1H-pyrazolo[4,3-c]pyridine

Hazard: Irritant

Formula: C₆H₄BrN₃
MW: 198.0200 g/mol
MDL Number: MFCD11518973
CAS Number: 633328-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR