4-(Methylsulfinylphenyl)boronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: (4-(Methylsulphinyl)phenyl)boronic acid

070038-1G 070038-25G
101943-044EA 466.15 USD
101943-044 101943-048
4-(Methylsulfinylphenyl)boronic acid ≥97%
4-(Methylsulfinylphenyl)boronic acid

Hazard: Irritant

Formula: C₇H₉BO₃S
MW: 184.02 g/mol
MDL Number: MFCD02093071
CAS Number: 166386-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR