(4,4-Difluorocyclohexyl)methanamine hydrochloride ≥97%
Supplier: Matrix Scientific
070033-1G
070033-25G
101943-020EA
961.54
USD
101943-020
101943-024
(4,4-Difluorocyclohexyl)methanamine hydrochloride ≥97%
(4,4-Difluorocyclohexyl)methanamine hydrochloride
Hazard: Irritant
Formula:
C₇H₁₄ClF₂N |
MDL Number:
MFCD13189562 CAS Number: 809273-65-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...