4-Methyl-1H-indazole-5-boronic acid ≥96%

Supplier: Matrix Scientific

Synonyms: (4-Methyl-1H-indazol-5-yl)boronic acid

070030-500MG 070030-5G
101943-004EA 349.23 USD
101943-004 101943-008
4-Methyl-1H-indazole-5-boronic acid ≥96%
4-Methyl-1H-indazole-5-boronic acid

Hazard: Irritant

Formula: C₈H₉BN₂O₂
MW: 175.9800 g/mol
MDL Number: MFCD12028644
CAS Number: 1245816-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR