2,4-Dichloro-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidine ≥95%

Supplier: Matrix Scientific

070019-500MG 070019-5G
101942-958EA 1106.15 USD
101942-958 101942-962
2,4-Dichloro-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidine ≥95%
2,4-Dichloro-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidine

Hazard: Irritant

Formula: C₇H₇Cl₂N₃
MDL Number: MFCD09835496
CAS Number: 1000578-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR