5,7-Dichloropyrazolo[1,5-a]pyrimidine ≥95%

Supplier: Matrix Scientific

069978-1G 069978-25G
101942-784EA 930.77 USD
101942-784 101942-788
5,7-Dichloropyrazolo[1,5-a]pyrimidine ≥95%
5,7-Dichloropyrazolo[1,5-a]pyrimidine

Hazard: Irritant

Formula: C₆H₃Cl₂N₃
MW: 188.0141 g/mol
MDL Number: MFCD08273918
CAS Number: 57489-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR