6-Chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
069961-250MG
069961-1G
101942-712EA
756.92
USD
101942-712
101942-716
6-Chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylic acid ≥95%
6-Chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylic acid
Hazard: Irritant
Formula:
C₈H₅ClN₂O₂ |
MDL Number:
MFCD09878690 CAS Number: 800402-07-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...