Isoquinoline-4-amine ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Aminoisoquinoline, Isoquinolin-4-amine
069890-1G
069890-25G
101942-408EA
58.46
USD
101942-408
101942-412
Isoquinoline-4-amine ≥97%
Isoquinoline-4-amine
Hazard: Irritant
Formula:
C₉H₈N₂ MW: 144.1700 g/mol |
MDL Number:
MFCD00034752 CAS Number: 23687-25-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...