4-Amino-6-chloro-7-azaindole ≥95%

Supplier: Matrix Scientific

069885-500MG 069885-5G
101942-382EA 989.23 USD
101942-382 101942-386
4-Amino-6-chloro-7-azaindole ≥95%
4-Amino-6-chloro-7-azaindole

Hazard: Irritant

Formula: C₇H₆ClN₃
MDL Number: MFCD09880161
CAS Number: 1000340-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR