1,2-Bis(epoxybutyl)carborane ≥98%

Supplier: Matrix Scientific

069871-250MG 069871-1G
101942-318EA 1396.92 USD
101942-318 101942-322
1,2-Bis(epoxybutyl)carborane ≥98%
1,2-Bis(epoxybutyl)carborane

Hazard: Irritant

Formula: C₁₀H₁₄B₁₀O₂
MDL Number: MFCD11114555
CAS Number: 28065-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR