2-Methyl-2H-indazole-5-boronic acid pinacol ester ≥96%

Supplier: Matrix Scientific

Synonyms: 2-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-indazole

069863-500MG 069863-5G
101942-282EA 407.69 USD
101942-282 101942-286
2-Methyl-2H-indazole-5-boronic acid pinacol ester ≥96%
2-Methyl-2H-indazole-5-boronic acid pinacol ester

Hazard: Irritant

Formula: C₁₄H₁₉BN₂O₂
MDL Number: MFCD11109436
CAS Number: 1189746-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR