2-Bromo-5-chloropyrazine ≥98%

Supplier: Matrix Scientific

069845-500MG 069845-5G
101942-204EA 300 USD
101942-204 101942-208
2-Bromo-5-chloropyrazine ≥98%
2-Bromo-5-chloropyrazine

Hazard: Irritant

Formula: C₄H₂BrClN₂
MW: 193.4291 g/mol
MDL Number: MFCD08460074
CAS Number: 912773-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR