2-Methyl-2H-indazole-5-carboxylic acid methyl ester ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2-methyl-2H-indazole-5-carboxylate

069841-250MG 069841-1G
101942-180EA 466.15 USD
101942-180 101942-184
2-Methyl-2H-indazole-5-carboxylic acid methyl ester ≥97%
2-Methyl-2H-indazole-5-carboxylic acid methyl ester

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂
MW: 190.2 g/mol
MDL Number: MFCD11109406
CAS Number: 1092351-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR