1-Methyl-1H-indazole-7-carboxylic acid methyl ester ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 1-methyl-1H-indazole-7-carboxylate

069839-500MG 069839-5G
101942-168EA 756.92 USD
101942-168 101942-172
1-Methyl-1H-indazole-7-carboxylic acid methyl ester ≥97%
1-Methyl-1H-indazole-7-carboxylic acid methyl ester

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂
MW: 190.2 g/mol
MDL Number: MFCD11109404
CAS Number: 1092351-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR