Methyl-1-methyl-1H-indazole-4-carboxylate ≥97%
Supplier: Matrix Scientific
Synonyms:
Methyl 1-methyl-1H-indazole-4-carboxylate
069835-500MG
069835-5G
101942-146EA
466.15
USD
101942-146
101942-150
Methyl-1-methyl-1H-indazole-4-carboxylate ≥97%
Methyl-1-methyl-1H-indazole-4-carboxylate
Hazard: Irritant
Formula:
C₁₀H₁₀N₂O₂ MW: 190.1986 g/mol |
MDL Number:
MFCD11109402 CAS Number: 1071428-42-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...