Methyl-1H-indazole-4-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 1H-indazole-4-carboxylate

069833-500MG 069833-5G
101942-134EA 20 USD
101942-134 101942-138
Methyl-1H-indazole-4-carboxylate ≥97%
Methyl 1H-indazole-4-carboxylate

Hazard: Irritant

Formula: C₉H₈N₂O₂
MW: 176.1700 g/mol
MDL Number: MFCD07371610
CAS Number: 192945-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR