3-Amino-6-fluoro-1-methyl-1H-indazole ≥98%
Supplier: Matrix Scientific
069827-500MG
069827-5G
101942-106EA
524.62
USD
101942-106
101942-110
3-Amino-6-fluoro-1-methyl-1H-indazole ≥98%
3-Amino-6-fluoro-1-methyl-1H-indazole
Hazard: Irritant
Formula:
C₈H₈FN₃ |
MDL Number:
MFCD11109398 CAS Number: 171809-13-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...