3-Amino-4-fluoro-1-methyl-1H-indazole ≥98%
Supplier: Matrix Scientific
Synonyms:
3-Amino-4-fluoro-1-methylindazole
069825-500MG
069825-5G
101942-094EA
384.62
USD
101942-094
101942-098
3-Amino-4-fluoro-1-methyl-1H-indazole ≥98%
3-Amino-4-fluoro-1-methyl-1H-indazole
Hazard: Irritant
Formula:
C₈H₈FN₃ MW: 165.1700 g/mol |
MDL Number:
MFCD11109397 CAS Number: 162502-44-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...