1H-Indazole-4-carboxylic acid ≥96%

Supplier: Matrix Scientific

069793-500MG 069793-5G
101941-958EA 395.38 USD
101941-958 101941-962
1H-Indazole-4-carboxylic acid ≥96%
1H-Indazole-4-carboxylic acid

Hazard: Irritant

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
MDL Number: MFCD03840644
CAS Number: 677306-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR