4-Amino-2-chloro-5-methylpyrimidine ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-5-methylpyrimidin-4-amine

069779-1G 069779-25G
101941-894EA 150.77 USD
101941-894 101941-898
4-Amino-2-chloro-5-methylpyrimidine ≥98%
4-Amino-2-chloro-5-methylpyrimidine

Hazard: Irritant

Formula: C₅H₆ClN₃
MW: 143.58 g/mol
MDL Number: MFCD02683249
CAS Number: 14394-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR