4,6-Dichloro-1H-pyrrolo[3,2-c]pyridine-2-carboxylic acid methyl ester

Supplier: Matrix Scientific

069774-500MG 069774-5G
101941-866EA 900.83 USD
101941-866 101941-870
4,6-Dichloro-1H-pyrrolo[3,2-c]pyridine-2-carboxylic acid methyl ester
4,6-Dichloro-1H-pyrrolo[3,2-c]pyridine-2-carboxylic acid methyl ester

Hazard: Irritant

Formula: C₉H₆Cl₂N₂O₂
MW: 245.07 g/mol
MDL Number: MFCD10699204
CAS Number: 871583-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR