6-Bromo-3-(methylthio)[1,2,4]triazolo[4,3-a]pyridine ≥98%
Supplier: Matrix Scientific
069767-500MG
069767-5G
101941-834EA
1047.69
USD
101941-834
101941-838
6-Bromo-3-(methylthio)[1,2,4]triazolo[4,3-a]pyridine ≥98%
6-Bromo-3-(methylthio)-[1,2,4]triazolo[4,3-a]pyridine
Hazard: Irritant
Formula:
C₇H₆BrN₃S |
MDL Number:
MFCD09475863 CAS Number: 941294-56-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...