2-Methyloxazole-4-carboxaldehyde ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Methyloxazole-4-carbaldehyde

069751-1G 069751-5G
101941-760EA 364.62 USD
101941-760 101941-762
2-Methyloxazole-4-carboxaldehyde ≥98%
2-Methyloxazole-4-carboxaldehyde

Hazard: Irritant

Formula: C₅H₅NO₂
MW: 111.1 g/mol
MDL Number: MFCD03265461
CAS Number: 113732-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR